Synthware™ Round Bottom Flasks, 4-Neck
With standard taper outer joints on both the center and vertical side necks
$231.82 - $850.35
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
31501308
|
Synthware™
F214250 |
25 mL | Center: 24/40; 24/40 side |
Each for $231.82
|
|
||||
31501309
|
Synthware™
F214500 |
500 mL | Center: 24/40; 24/40 side |
Each for $239.74
|
|
||||
31501310
|
Synthware™
F212500 |
500 mL | Center: 29/42; 24/40 side |
Each for $242.21
|
|
||||
31501311
|
Synthware™
F218500 |
500 mL | 29/42 |
Each for $245.09
|
|
||||
31501312
|
Synthware™
F214000 |
1000 mL | Center: 24/40; 24/40 side |
Each for $295.29
|
|
||||
31501313
|
Synthware™
F21841L |
1000 mL | Center: 29/42; 24/40 side |
Each for $298.38
|
|
||||
31501314
|
Synthware™
F21842L |
2000 mL | Center: 29/42; 24/40 side |
Each for $339.15
|
|
||||
31501315
|
Synthware™
F21342L |
2000 mL | Center: 34/45; 24/40 side |
Each for $348.41
|
|
||||
31502231
|
Synthware™
F21543L |
3 L | Center: 45/50; Side: 24/40 |
Each for $597.74
|
|
||||
31502232
|
Synthware™
F21583L |
3 L | Center: 45/50; Side: 29/42 |
Each for $606.97
|
|
||||
31502233
|
Synthware™
F21845L |
5 L | Center: 29/42; Side: 24/40 |
Each for $786.68
|
|
||||
31502234
|
Synthware™
F21545L |
5 L | Center: 45/50; Side: 24/40 |
Each for $842.12
|
|
||||
31502235
|
Synthware™
F21585L |
5 L | Center: 45/50; Side: 29/42 |
Each for $850.35
|
|
||||
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Round bottom flask, 4-Neck |