Synthware™ Schlenk Flasks, Modified, with Glass Stopcock
Kjeldahl shape to facilitate heating and stirring
Supplier:  Synthware™ F901410G
 View more versions of this product
Catalog No. 31501355
Explore available promotions
Description
Description
  • Provides easy transfer of liquids and solids
  • Glass stopcock with 2 mm bore
  • Top with standard taper outer joint
Specifications
Specifications
Schlenk Flask, Modified
Borosilicate Glass (3.3)
Autoclavable
No
Non-sterile
Round
10 mL
10 mL
14/20
Reusable
Rimmed
A
Stopper
14/20