Synthware™ Schlenk Flasks, Modified, with High Vacuum Valve
Kjeldahl shape to facilitate heating and stirring
Supplier:  Synthware™ F901200H
 View more versions of this product
Catalog No. 31501374
Explore available promotions
Description
Description
  • Provides easy transfer of liquids and solids
  • With 0-4 mm high vacuum valve
  • Top with standard taper outer joint
Specifications
Specifications
Schlenk Flask, Modified, with High Vacuum Valve
Borosilicate Glass (3.3)
Autoclavable
No
Non-sterile
Round
200 mL
200 mL
14/20
Reusable
Rimmed
A
Stopper
14/20