Synthware™ Schlenk Flasks, Modified, with High Vacuum Valve
Kjeldahl shape to facilitate heating and stirring
$137.44 - $195.38
Specifications
Description | Schlenk Flask, Modified, with High Vacuum Valve |
---|---|
Autoclavable | Autoclavable |
Mouth Style | Rimmed |
Sterility | Non-sterile |
Class | A |
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
31501367
|
Synthware™
F901425H |
25 mL | 14/20 |
Each for $137.44
|
|
||||
31501368
|
Synthware™
F901450H |
50 mL | 14/20 |
Each for $140.79
|
|
||||
31501370
|
Synthware™
F902450H |
50 mL | 24/40 |
Each for $144.74
|
|
||||
31501369
|
Synthware™
F901950H |
50 mL | 19/22 |
Each for $143.65
|
|
||||
31501372
|
Synthware™
F909100H |
100 mL | 19/22 |
Each for $151.53
|
|
||||
31501371
|
Synthware™
F901100H |
100 mL | 14/20 |
Each for $145.76
|
|
||||
31501374
|
Synthware™
F901200H |
200 mL | 14/20 |
Each for $163.53
|
|
||||
31501373
|
Synthware™
F904100H |
100 mL | 24/40 |
Each for $155.09
|
|
||||
31501375
|
Synthware™
F909200H |
200 mL | 19/22 |
Each for $165.12
|
|
||||
31501376
|
Synthware™
F904200H |
200 mL | 24/40 |
Each for $167.88
|
|
||||
31501377
|
Synthware™
F904500H |
500 mL | 24/40 |
Each for $171.71
|
|
||||
31501378
|
Synthware™
F904000H |
1000 mL | 24/40 |
Each for $195.38
|
|
||||
Description
- Provides easy transfer of liquids and solids
- With 0-4 mm high vacuum valve
- Top with standard taper outer joint
Specifications
Schlenk Flask, Modified, with High Vacuum Valve | |
Rimmed | |
A | |
Stopper |
Autoclavable | |
Non-sterile | |
Round |