Synthware™ Squibb Separatory Funnels with Glass Stopcock
Supplied complete with flask length glass stopper
Supplier: Synthware™ F780030
Description
Synthware Squibb Separatory Funnel is used to separate two immiscible liquids of different densities.Specifications
30 mL | |
Clear | |
1 | |
Glass | |
Squibb Separatory Funnel |
Glass | |
Organic Synthesis | |
2mm | |
13 |