missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Squibb Separatory Funnels with Glass Stopcock

Supplied complete with flask length glass stopper

$107.41 - $280.32

Specifications

Material Glass
Color Clear
For Use With (Application) Organic Synthesis
Quantity 1
Stopper Material Glass
View More Specs

Products 6
Catalog Number Mfr. No. Capacity (Metric) Stopper Number Stopcock Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Stopper Number Stopcock Price Quantity  
31501681
View Documents
Synthware™
F780030
30 mL 13 2mm
Each for $107.41
 
31501684
View Documents
Synthware™
F780250
250 mL 22 4mm
Each for $155.18
 
31501682
View Documents
Synthware™
F780060
60 mL 16 2mm
Each for $113.62
 
31501683
View Documents
Synthware™
F780125
125 mL 22 2mm
Each for $134.35
 
31501685
View Documents
Synthware™
F780500
500 mL 27 4mm
Each for $188.12
 
31501686
View Documents
Synthware™
F787000
1000 mL 27 4mm
Each for $280.32
 
Description

Description

Synthware Squibb Separatory Funnel is used to separate two immiscible liquids of different densities.
Specifications

Specifications

Glass
Organic Synthesis
Glass
Clear
1
Squibb Separatory Funnel
SDS
missing translation for 'documents'

missing translation for 'documents'