Synthware™ Storage Flasks with Side Arm
Supplied with high vacuum valve with PTFE O-ring at tip
Supplier:  Synthware™ F464500
 View more versions of this product
Catalog No. 31501396
Explore available promotions
Description
Description
SYNTHWARE storage flask with standard taper outer joint and high vacuum valve. Valve has a PTFE O-ring at the tip. Valve is used to control liquid or gas flow into the flask.
Specifications
Specifications
Clear
500 mL
Organic Synthesis
Valve Size: 0-4mm
Borosilicate Glass (3.3)
24/40