Synthware™ Three Neck Round Bottom Flasks, Vertical Side Neck with Bottom Outlet
With standard taper outer joints on both center and vertical side necks
Supplier: Synthware™ F42441L
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
- 4 mm straight bore PTFE stopcock lower outlet
Specifications
Round Bottom Flask, 3-Neck Bottom Outlet | |
1000 mL | |
Center: 24/40; 24/40 side |