missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Three Neck Round Bottom Flasks, Vertical Side Neck with Bottom Outlet

With standard taper outer joints on both center and vertical side necks

$565.13 - $753.73

Specifications

Description Round Bottom Flask, 3-Neck Bottom Outlet
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity  
31501300
View Documents
Synthware™
F424450
500 mL Center: 24/40; 24/40 side
Each for $565.13
Only null left
 
31501301
View Documents
Synthware™
F42441L
1000 mL Center: 24/40; 24/40 side
Each for $627.67
Only null left
 
31501302
View Documents
Synthware™
F42442L
2000 mL Center: 24/40; 24/40 side
Each for $710.10
Only null left
 
31501303
View Documents
Synthware™
F42542L
2000 mL Center: 45/50; 24/40 side
Each for $753.73
Only null left
 
Description

Description

  • All sizes are hand blown from tubing to ensure uniform wall thickness
  • 4 mm straight bore PTFE stopcock lower outlet
Specifications

Specifications

Round Bottom Flask, 3-Neck Bottom Outlet
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'