Synthware™ Two Neck Round Bottom Flask, Angled Side Neck
Two Neck Round Bottom Flask with standard taper outer joints on both center and vertical side necks.
Supplier:  Synthware™ F414150
 View more versions of this product
Catalog No. 31501213
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Two Neck Round Bottom Flasks, Angled Side Neck
Borosilicate Glass (3.3)
Center: 24/40, Side: 14/20
Reusable
Beaded
Round
50 mL
Two-neck Flask
Autoclavable
No
Non-sterile