Synthware™ Two Neck Round Bottom Flask, Angled Side Neck
Two Neck Round Bottom Flask with standard taper outer joints on both center and vertical side necks.
$78.71 - $480.32
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Product Type | Two-neck Flask |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | No |
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
31501207
|
Synthware™
F411110 |
10 mL | Center: 14/20, Side: 14/20 |
Each for $78.71
|
|
||||
31501210
|
Synthware™
F419925 |
25 mL | Center: 19/22, Side: 19/22 |
Each for $82.53
|
|
||||
31501209
|
Synthware™
F411925 |
25 mL | Center: 19/22, Side: 14/20 |
Each for $82.24
|
|
||||
31501211
|
Synthware™
F411150 |
50 mL | Center: 14/20, Side: 14/20 |
Each for $82.94
|
|
||||
31501208
|
Synthware™
F411125 |
25 mL | Center: 14/20, Side: 14/20 |
Each for $81.82
|
|
||||
31501212
|
Synthware™
F419950 |
50 mL | Center: 19/22, Side: 19/22 |
Each for $83.21
|
|
||||
31501213
|
Synthware™
F414150 |
50 mL | Center: 24/40, Side: 14/20 |
Each for $85.94
|
|
||||
31501214
|
Synthware™
F414450 |
50 mL | Center: 24/40, Side: 24/40 |
Each for $87.06
|
|
||||
31501215
|
Synthware™
F411100 |
100 mL | Center: 14/20, Side: 14/20 |
Each for $84.32
|
|
||||
31501216
|
Synthware™
F411900 |
100 mL | Center: 19/22, Side: 14/20 |
Each for $84.79
|
|
||||
31501217
|
Synthware™
F419900 |
100 mL | Center: 19/22, Side: 19/22 |
Each for $84.94
|
|
||||
31501218
|
Synthware™
F414100 |
100 mL | Center: 24/40, Side: 14/20 |
Each for $87.21
|
|
||||
31501219
|
Synthware™
F419100 |
100 mL | Center: 24/40, Side: 19/22 |
Each for $104.50
|
|
||||
31501220
|
Synthware™
F414400 |
100 mL | Center: 24/40, Side: 24/40 |
Each for $114.24
|
|
||||
31501222
|
Synthware™
F410250 |
250 mL | Center: 19/22, Side: 14/20 |
Each for $89.85
|
|
||||
31501221
|
Synthware™
F411250 |
250 mL | Center: 14/20, Side: 14/20 |
Each for $88.35
|
|
||||
31501223
|
Synthware™
F419250 |
250 mL | Center: 19/22, Side: 19/22 |
Each for $93.41
|
|
||||
31501224
|
Synthware™
F414251 |
250 mL | Center: 24/40, Side: 14/20 |
Each for $111.74
|
|
||||
31501225
|
Synthware™
F414259 |
250 mL | Center: 24/40, Side: 19/22 |
Each for $112.50
|
|
||||
31501226
|
Synthware™
F414250 |
250 mL | Center: 24/40, Side: 24/40 |
Each for $118.03
|
|
||||
31501227
|
Synthware™
F411501 |
500 mL | Center: 19/22, Side: 14/20 |
Each for $119.97
|
|
||||
31501228
|
Synthware™
F414501 |
500 mL | Center: 24/40, Side: 14/20 |
Each for $123.74
|
|
||||
31501229
|
Synthware™
F414504 |
500 mL | Center: 24/40, Side: 24/40 |
Each for $125.74
|
|
||||
31501230
|
Synthware™
F415501 |
500 mL | Center: 29/42, Side: 24/40 |
Each for $137.97
|
|
||||
31501231
|
Synthware™
F415502 |
500 mL | Center: 29/42, Side: 29/42 |
Each for $146.59
|
|
||||
31501232
|
Synthware™
F41611L |
1000 mL | Center: 24/40, Side: 14/20 |
Each for $146.68
|
|
||||
31501233
|
Synthware™
F41621L |
1000 mL | Center: 24/40, Side: 19/22 |
Each for $147.38
|
|
||||
31501234
|
Synthware™
F41241L |
1000 mL | Center: 24/40, Side: 24/40 |
Each for $152.38
|
|
||||
31501235
|
Synthware™
F41641L |
1000 mL | Center: 29/42, Side: 29/42 |
Each for $172.76
|
|
||||
31502184
|
Synthware™
F41242L |
2 L | Center: 24/40, Side: 24/40 |
Each for $287.06
|
|
||||
31502185
|
Synthware™
F41712L |
2 L | Center: 29/42, Side: 24/40 |
Each for $290.71
|
|
||||
31502186
|
Synthware™
F41722L |
2 L | Center: 29/42, Side: 29/42 |
Each for $293.50
|
|
||||
31502187
|
Synthware™
F41732L |
2 L | Center: 34/45, Side: 24/40 |
Each for $299.06
|
|
||||
31502188
|
Synthware™
F41742L |
2 L | Center: 34/45, Side: 29/42 |
Each for $299.06
|
|
||||
31502189
|
Synthware™
F41752L |
2 L | Center: 45/50, Side: 24/40 |
Each for $305.56
|
|
||||
31502190
|
Synthware™
F41762L |
2 L | Center: 45/50, Side: 29/42 |
Each for $305.56
|
|
||||
31502191
|
Synthware™
F41813L |
3 L | Center: 29/42, Side: 24/40 |
Each for $373.32
|
|
||||
31502192
|
Synthware™
F41823L |
3 L | Center: 29/42, Side: 29/42 |
Each for $380.82
|
|
||||
31502193
|
Synthware™
F41833L |
3 L | Center: 34/45, Side: 24/40 |
Each for $388.35
|
|
||||
31502194
|
Synthware™
F41843L |
3 L | Center: 34/45, Side: 29/42 |
Each for $395.85
|
|
||||
31502195
|
Synthware™
F41853L |
3 L | Center: 45/50, Side: 24/40 |
Each for $402.50
|
|
||||
31502196
|
Synthware™
F41863L |
3 L | Center: 45/50, Side: 29/42 |
Each for $408.32
|
|
||||
31502197
|
Synthware™
F41915L |
5 L | Center: 29/42, Side: 24/40 |
Each for $436.38
|
|
||||
31502198
|
Synthware™
F41925L |
5 L | Center: 29/42, Side: 29/42 |
Each for $447.00
|
|
||||
31502199
|
Synthware™
F41935L |
5 L | Center: 34/45, Side: 24/40 |
Each for $455.29
|
|
||||
31502200
|
Synthware™
F41945L |
5 L | Center: 34/45, Side: 29/42 |
Each for $464.38
|
|
||||
31502201
|
Synthware™
F41955L |
5 L | Center: 45/50, Side: 24/40 |
Each for $471.24
|
|
||||
31502202
|
Synthware™
F41965L |
5 L | Center: 45/50, Side: 29/42 |
Each for $480.32
|
|
||||
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Borosilicate Glass (3.3) | |
Autoclavable | |
No | |
Non-sterile |
Two-neck Flask | |
Reusable | |
Beaded | |
Round |