Synthware™ Two Neck Round Bottom Flask, Angled Side Neck
Two Neck Round Bottom Flask with standard taper outer joints on both center and vertical side necks.
Supplier:  Synthware™ F41732L
 View more versions of this product
Catalog No. 31502187
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Clear
Borosilicate Glass (3.3)
Two-neck Flask
Autoclavable
No
Non-sterile
2 L
Organic Synthesis
Center: 34/45, Side: 24/40
Reusable
Beaded
Round