Innovatek Syphilis RPR Antigen Latex Test Kits and Controls
Innovatek Syphilis RPR Antigen Latex Test Kits and Controls provide results within 3 minutes
Supplier:  Innovatek 419110605
 View more versions of this product
Catalog No. 361044799
Explore available promotions
Description
Description
  • Fast 2 to 3 minute rotation
  • Complete kit includes all reagents and controls
  • Disposable test cards
  • 6 to 12 months of shelf life
  • Sensitive and specific
Specifications
Specifications
Syphilis RPR Antigen
Single-use
≈3 min.
Latex Test Kit
500 Tests/Kit