missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Innovatek Syphilis RPR Antigen Latex Test Kits and Controls
Innovatek Syphilis RPR Antigen Latex Test Kits and Controls provide results within 3 minutes
$311.21 - $734.82
Specifications
Detectable Analytes | Syphilis RPR Antigen |
---|---|
Disposable | Single-use |
Test Time | ≈3 min. |
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
361045043
|
Innovatek
419110603 |
Latex Test Kit | 100 Tests/Kit |
Pack of 100 Each for $311.21
|
|
||||
361044799
|
Innovatek
419110605 |
Latex Test Kit | 500 Tests/Kit |
Pack of 500 Each for $734.82
|
|
||||
361045044
|
Innovatek
419110655 |
Control Serum Set | 3 Vials/Set |
Pack of 3 Each for $318.42
|
|
||||
Description
- Fast 2 to 3 minute rotation
- Complete kit includes all reagents and controls
- Disposable test cards
- 6 to 12 months of shelf life
- Sensitive and specific
Specifications
Syphilis RPR Antigen | |
Single-use | |
≈3 min. |