missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Syringaldehyde, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1293306

 View more versions of this product

Catalog No. AAA1293306


Explore available promotions

Description

Description

Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

Solubility
Very sparingly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to air. Incompatible with strong oxidizing agents, and bases.
Specifications

Specifications

Syringaldehyde
110°C to 113°C
C9H10O4
5 g
Air Sensitive
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
KCDXJAYRVLXPFO-UHFFFAOYSA-N
4-hydroxy-3,5-dimethoxybenzaldehyde
8655
182.18
134-96-3
192°C to 193°C (14 mmHg)
MFCD00006943
784514
14,9015
Very sparingly soluble in water.
COC1=CC(C=O)=CC(OC)=C1O
182.18
CHEBI:67380
≥98%

Chemical Identifiers

134-96-3
182.18
KCDXJAYRVLXPFO-UHFFFAOYSA-N
8655
4-hydroxy-3,5-dimethoxybenzaldehyde
C9H10O4
MFCD00006943
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
CHEBI:67380
COC1=CC(C=O)=CC(OC)=C1O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

EINECSNumber : 205-167-5

RTECSNumber : CU5760000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only