Learn More
Syringaldehyde, 98+%, Thermo Scientific Chemicals
$57.15 - $399.93
Chemical Identifiers
CAS | 134-96-3 |
---|---|
Molecular Formula | C9H10O4 |
Molecular Weight (g/mol) | 182.18 |
MDL Number | MFCD00006943 |
InChI Key | KCDXJAYRVLXPFO-UHFFFAOYSA-N |
Synonym | syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde |
PubChem CID | 8655 |
ChEBI | CHEBI:67380 |
IUPAC Name | 4-hydroxy-3,5-dimethoxybenzaldehyde |
SMILES | COC1=CC(C=O)=CC(OC)=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1293306
|
Thermo Scientific Chemicals
A1293306 |
5 g |
Each for $57.15
|
|
||||||
AAA1293314
|
Thermo Scientific Chemicals
A1293314 |
25 g |
Each for $113.77
|
|
||||||
AAA1293322
|
Thermo Scientific Chemicals
A1293322 |
100 g |
Each for $399.93
|
|
||||||
Description
Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSyringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.
Solubility
Very sparingly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to air. Incompatible with strong oxidizing agents, and bases.
Chemical Identifiers
134-96-3 | |
182.18 | |
KCDXJAYRVLXPFO-UHFFFAOYSA-N | |
8655 | |
4-hydroxy-3,5-dimethoxybenzaldehyde |
C9H10O4 | |
MFCD00006943 | |
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde | |
CHEBI:67380 | |
COC1=CC(C=O)=CC(OC)=C1O |
Specifications
134-96-3 | |
192°C to 193°C (14 mmHg) | |
MFCD00006943 | |
784514 | |
14,9015 | |
Very sparingly soluble in water. | |
COC1=CC(C=O)=CC(OC)=C1O | |
182.18 | |
CHEBI:67380 | |
≥98% |
110°C to 113°C | |
C9H10O4 | |
5 g | |
Air Sensitive | |
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde | |
KCDXJAYRVLXPFO-UHFFFAOYSA-N | |
4-hydroxy-3,5-dimethoxybenzaldehyde | |
8655 | |
182.18 | |
Syringaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 205-167-5
RTECSNumber : CU5760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only