Sartorius Tacta™ Mechanical Pipet Multipacks
Multipacks combine a set of Tacta single-channel mechanical pipets with matching tip trays and useful accessories. Sartorius Tacta Mechanical Pipet Multipacks provides best alternative to replace your used mechanical pipets.
Supplier:  Sartorius LH729670
 View more versions of this product
Catalog No. 14560030
Explore available promotions
Includes: 3 x Single-channel pipets (0.5 to 10μL, 10 to 100μL, 100 to 1000μL), Tip Racks of 96 tips (0.2 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand, 3 x Single Pipet Holders
Description
Description
  • Comfortable to hold due to ergonomically designed handle
  • Low pipetting and tip ejection forces
  • Smooth tip ejection with the new Optiject™ technology
  • Unique Optilock™ system provides flexibility for volume adjustment and locking
  • Calibration adjustment to provide accurate results for various liquid types
  • Reliable results, even over lengthy pipetting periods
  • Easy-to-read, four-digit volume display helps to set exact volumes
  • Easy to clean with only three parts to disassemble
  • Fully autoclavable
  • High-quality standard Optifit tips for various applications
Warranty and Services
Two years
Specifications
Specifications
Autoclavable
1
Mechanical Pipette Multipacks
Four-digit volume display
10/Pk.
2 Years
79 g (1-channel, 1000μL); 138 g (8-channel, 300μL)
Tacta
Tacta
0.5 to 10, 10 to 100, 100 to 1000 μL
3 x Single-channel pipets (0.5 to 10μL, 10 to 100μL, 100 to 1000μL), Tip Racks of 96 tips (0.2 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand, 3 x Single Pipet Holders
High
138 mm (1-channel, 1000μL); 240 mm (8-channel, 300μL)