Sartorius Tacta™ Mechanical Pipet Multipacks
Multipacks combine a set of Tacta single-channel mechanical pipets with matching tip trays and useful accessories. Sartorius Tacta Mechanical Pipet Multipacks provides best alternative to replace your used mechanical pipets.
$1542.15 - $2649.02
Specifications
Autoclavable Autoclavable
Model Tacta
Product Line Tacta
Type Mechanical Pipette Multipacks
Display Four-digit volume display
View More Specs
Products 8
Catalog Number Mfr. No. Type Quantity Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Type Quantity Volume (Metric) Price Quantity  
14560033
View Documents Promotion Details
Sartorius
LH729673
Mechanical Pipette Multipacks 20/Pk. 0.5 to 10, 10 to 100, 20 to 200, 100 to 1000 μL
Each for $1,985.64
14560034
View Documents Promotion Details
Sartorius
LH729674
Mechanical Pipette Multipacks 10/Pk. 0.5 to 10, 10 to 100, 20 to 200, 100 to 1000, 500 to 5000 μL
Each for $2,288.35
14560030
View Documents Promotion Details
Sartorius
LH729670
Mechanical Pipette Multipacks 10/Pk. 0.5 to 10, 10 to 100, 100 to 1000 μL
Each for $1,542.15
14560032
View Documents Promotion Details
Sartorius
LH729672
Mechanical Pipette Multipacks 10/Pk. 0.5 to 10, 2 to 20, 20 to 200, 100 to 1000 μL
Each for $1,952.55
14560031
View Documents Promotion Details
Sartorius
LH729671
Mechanical Pipette Multipacks 20/Pk.
Each for $1,542.15
14560036
View Documents Promotion Details
Sartorius
LH729676
Mechanical Pipette Multipacks 1/Pk. 0.5 to 10, 20 to 200, 100 to 1000, 30 to 300 μL
Each for $2,649.02
14560035
View Documents Promotion Details
Sartorius
LH729675
Mechanical Pipette Multipacks 20/Pk. 2 to 20, 10 to 100, 20 to 200, 100 to 1000, 500 to 5000 μL
Each for $2,288.35
14560037
View Documents Promotion Details
Sartorius
LH729677
Mechanical Pipette Multipacks 1/Pk. 100 to 1000, 500 to 5000 μL, 1 to 10 mL
Each for $1,554.58
Description
Description
  • Comfortable to hold due to ergonomically designed handle
  • Low pipetting and tip ejection forces
  • Smooth tip ejection with the new Optiject™ technology
  • Unique Optilock™ system provides flexibility for volume adjustment and locking
  • Calibration adjustment to provide accurate results for various liquid types
  • Reliable results, even over lengthy pipetting periods
  • Easy-to-read, four-digit volume display helps to set exact volumes
  • Easy to clean with only three parts to disassemble
  • Fully autoclavable
  • High-quality standard Optifit tips for various applications
Warranty
Two years
Specifications
Specifications
Autoclavable
Tacta
Four-digit volume display
2 Years
79 g (1-channel, 1000μL); 138 g (8-channel, 300μL)
Tacta
Mechanical Pipette Multipacks
High
138 mm (1-channel, 1000μL); 240 mm (8-channel, 300μL)
SDS
missing translation for 'documents'
missing translation for 'documents'