missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Tacta™ Mechanical Pipet Multipacks

Multipacks combine a set of Tacta single-channel mechanical pipets with matching tip trays and useful accessories. Sartorius Tacta Mechanical Pipet Multipacks provides best alternative to replace your used mechanical pipets.

Supplier:  Sartorius LH729677

 View more versions of this product

Catalog No. 14560037


Explore available promotions

Includes: 3 MAX Single-channel pipets (100 to 1000μL, 500 to 5000μL, 1 to 10mL), Tip Racks to 96 tips (10 to 1000μL) and Bulk in a bag: 0.1 to 5mL (100 tips), 1 to 10mL (250 tips), 1 x Linear Stand, 3 x Pipet Holders

Description

Description

  • Comfortable to hold due to ergonomically designed handle
  • Low pipetting and tip ejection forces
  • Smooth tip ejection with the new Optiject™ technology
  • Unique Optilock™ system provides flexibility for volume adjustment and locking
  • Calibration adjustment to provide accurate results for various liquid types
  • Reliable results, even over lengthy pipetting periods
  • Easy-to-read, four-digit volume display helps to set exact volumes
  • Easy to clean with only three parts to disassemble
  • Fully autoclavable
  • High-quality standard Optifit tips for various applications

Warranty

Two years

Specifications

Specifications

Autoclavable
1
Mechanical Pipette Multipacks
Four-digit volume display
1/Pk.
2 Years
79 g (1-channel, 1000μL); 138 g (8-channel, 300μL)
Tacta
Tacta
100 to 1000, 500 to 5000 μL, 1 to 10 mL
3 MAX Single-channel pipets (100 to 1000μL, 500 to 5000μL, 1 to 10mL), Tip Racks to 96 tips (10 to 1000μL) and Bulk in a bag: 0.1 to 5mL (100 tips), 1 to 10mL (250 tips), 1 x Linear Stand, 3 x Pipet Holders
High
138 mm (1-channel, 1000μL); 240 mm (8-channel, 300μL)
SDS
Documents

Documents

Promotions

Promotions