missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tartaric Acid, 10% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 837132
Specifications
Tartaric Acid | |
0.89,0.0891 | |
0.93,0.0928 | |
MFCD00064207 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
10% (w/v) | |
150.09 | |
CHEBI:15671 | |
Natural Poly Bottle |
87-69-4,7732-18-5 | |
Colorless to Yellow | |
C4H6O6 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid | |
OC(C(O)C(O)=O)C(O)=O | |
2,3-dihydroxybutanedioic acid | |
444305 | |
1 L | |
Liquid |
Chemical Identifiers
87-69-4 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
444305 | |
2,3-dihydroxybutanedioic acid |
C4H6O6 | |
MFCD00064207 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid | |
CHEBI:15671 | |
OC(C(O)C(O)=O)C(O)=O |