Tecniplast™ Stainless Steel Wire Cage Lids and Feeders
Secure and easty to handle
Supplier:  Tecniplast™ 1144B116
 View more versions of this product
Catalog No. 0128821
Explore available promotions
Description
Description
  • U-shaped hopper allows easy access to food and won't entrap food or contaminants
  • Cross-wire members prevent bending
  • 0.07cm wire spacing
  • Folding hopper divider allows lids to be stacked for storage
  • Central divider prevents animal from crawling into feeder
  • Can be used with or without a raised wire floor
  • Autoclavable
Specifications
Specifications
Wire Cage Lid with Feeder
Mice
0.07 cm
Stainless Steel
Cage size B