missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Temperature Chart Recorders

Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.

Supplier:  Thermo Scientific™ 63837A

 Afficher plus de versions de ce produit

Numéro de catalogue. 36-101-7623


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.

Features:
  • Seven-day chart recorders record temperatures from -100° to 0°C (-40° to +38° on 6183-7)
  • 6 in. (15.2cm) chart
  • Use pressure-sensitive paper, require no inking
  • Freestanding model is customer installed and must be installed by a qualified professional
  • Panel-mounted model is factory installed; order when purchasing refrigerator or freezer

Ordering Information:

Available in range of voltages; specify voltage when ordering.

Includes:

Pack of 50 charts

Spécifications

Spécifications

Seven-day chart recorders
-40°C to +25°C
General-purpose, Chromatography Refrigerators
6 in.
Recorder
Freestanding
Freestanding
7 Days
15.2 cm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions