missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Temperature Chart Recorders

Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.

$5,115.70 - $5,115.70

Specifications

Description Seven-day chart recorders
Static Holding Time 7 Days
Diameter (English) 6 in.
Diameter (Metric) 15.2 cm
Monitoring Options Recorder
View More Specs

Products 1
Catalog Number Mfr. No. Temperature Range Type For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Temperature Range Type For Use With (Equipment) Price Quantity  
36-101-7623
View Documents
Thermo Scientific™
63837A
-40°C to +25°C Freestanding General-Purpose, Chromatography Refrigerators N/A
 
Description

Description

Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.

Features:
  • Seven-day chart recorders record temperatures from -100° to 0°C (-40° to +38° on 6183-7)
  • 6 in. (15.2cm) chart
  • Use pressure-sensitive paper, require no inking
  • Freestanding model is customer installed and must be installed by a qualified professional
  • Panel-mounted model is factory installed; order when purchasing refrigerator or freezer

Ordering Information:

Available in range of voltages; specify voltage when ordering.

Includes:

Pack of 50 charts

Specifications

Specifications

Seven-day chart recorders
6 in.
Recorder
7 Days
15.2 cm
SDS
missing translation for 'documents'

missing translation for 'documents'