DuPont™ Tempro™ TM 127S Coveralls
Offer an extra layer of protection for primary fire-resistant clothing
Supplier:  DuPont™ TM127SBU5X002500
 View more versions of this product
Catalog No. 19-100-386
Explore available promotions
Alert:
Should not be worn in presence of hazardous materials.
Description
Description
Worn over flame-resistant clothing to provide a barrier to nonhazardous particles and aerosols. Fit easily over fire-resistant clothing.
  • Elastic wrists and ankles
  • Attached hood with elastic around face
  • Front zipper closure
  • Serged seam
  • Nonwoven wood-pulp/polyester, flame-retardant, liquid repellant fabric
  • Protect and preserve primary flame-resistant clothing
  • Disposable
Can be used in steel mills, coal mines, maintenance, refineries, utilities, laboratories
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
Specifications
5X-Large
Blue
Elastic
Hooded
Zipper
Non-woven Wood Pulp, Polyester Fabric
33.5 in.
Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities
102.87 cm
6.7 to 6.10 ft.
Tempro™
Single-use
Elastic
Standard
Serged
53.75 to 57.25 in.
136.52 to 145.41 cm
40.5 in.
33.5 in.
2.04 to 1.85 m