missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tempro™ TM 127S Coveralls
Offer an extra layer of protection for primary fire-resistant clothing
$290.84 - $444.57
Specifications
Material | Tempro™ |
---|---|
Color | Blue |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Alert:
Should not be worn in presence of hazardous materials.
Catalog Number | Mfr. No. | Size | Height (English) | Height (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Height (English) | Height (Metric) | Price | Quantity | |||
19-175-272
|
DuPont™
TM127SBU6X002500 |
6X-Large | 6.9 to 7.1 ft. | 2.1 to 2.16 m |
Case of 25 Each for $411.58
|
|
|||
19-100-386
|
DuPont™
TM127SBU5X002500 |
5X-Large | 6.7 to 6.10 ft. | 2.04 to 1.85 m |
Case of 25 Each for $381.13
|
|
|||
29-635-9904
|
DuPont™
TM127SBU4X002500 |
4X-Large | 6.4 to 6.7 ft. | 1.95 to 2.04 m |
Case of 25 Each for $352.87
|
|
|||
19-130-7801
|
DuPont™
TM127SBUXL002500 |
X-Large | 5.8 to 6.2 ft. | 1.76 to 1.88 m |
Case of 25 Each for $315.71
|
|
|||
19-130-7800
|
DuPont™
TM127SBULG002500 |
Large | 5.5 to 5.9 ft. | 1.67 to 1.8 m |
Case of 25 Each for $290.84
|
|
|||
19-130-7803
|
DuPont™
TM127SBU3X002500 |
3X-Large | 6.2 to 6.4 ft. | 1.88 to 1.95 m |
Case of 25 Each for $326.75
|
|
|||
29-663-156
|
DuPont™
TM127SBU2X002500 |
2X-Large | 6 to 6.4 ft. | 1.82 to 1.95 m |
Case of 25 Each for $339.25
|
|
|||
29-663-153
|
DuPont™
TM127SBUMD002500 |
Medium | 5.3 to 5.7 ft. | 1.6 to 1.73 m |
Case of 25 Each for $290.84
|
|
|||
19-175-273
|
DuPont™
TM127SBU7X002500 |
7X-Large | 7 to 7.4 ft. | 2.13 to 2.25 m |
Case of 25 Each for $444.57
|
|
|||
Description
Worn over flame-resistant clothing to provide a barrier to nonhazardous particles and aerosols. Fit easily over fire-resistant clothing.
- Elastic wrists and ankles
- Attached hood with elastic around face
- Front zipper closure
- Serged seam
- Nonwoven wood-pulp/polyester, flame-retardant, liquid repellant fabric
- Protect and preserve primary flame-resistant clothing
- Disposable
Can be used in steel mills, coal mines, maintenance, refineries, utilities, laboratories
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
Tempro™ | |
Elastic | |
Hooded | |
Zipper | |
Non-woven Wood Pulp, Polyester Fabric |
Blue | |
Elastic | |
Standard | |
Serged | |
Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities |