DuPont™ Tempro™ TM 127S Coveralls
Offer an extra layer of protection for primary fire-resistant clothing
Supplier:  DuPont™ TM127SBU3X002500
 View more versions of this product
Catalog No. 19-130-7803
Explore available promotions
Alert:
Should not be worn in presence of hazardous materials.
Description
Description
Worn over flame-resistant clothing to provide a barrier to nonhazardous particles and aerosols. Fit easily over fire-resistant clothing.
  • Elastic wrists and ankles
  • Attached hood with elastic around face
  • Front zipper closure
  • Serged seam
  • Nonwoven wood-pulp/polyester, flame-retardant, liquid repellant fabric
  • Protect and preserve primary flame-resistant clothing
  • Disposable
Can be used in steel mills, coal mines, maintenance, refineries, utilities, laboratories
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
Specifications
3X-Large
Blue
Elastic
Standard
Serged
47.25 to 50.75 in.
120 to 128.9 cm
Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities
97.79 cm
6.2 to 6.4 ft.
Tempro™
Elastic
Hooded
Zipper
Non-woven Wood Pulp, Polyester Fabric
31.5 in.
80 cm
38.5 in.
30.25 in.
1.88 to 1.95 m