missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tempro™ TM 127S Coveralls
Offer an extra layer of protection for primary fire-resistant clothing
Supplier: DuPont™ TM127SBUMD002500
Alert:
Should not be worn in presence of hazardous materials.
Description
Worn over flame-resistant clothing to provide a barrier to nonhazardous particles and aerosols. Fit easily over fire-resistant clothing.
- Elastic wrists and ankles
- Attached hood with elastic around face
- Front zipper closure
- Serged seam
- Nonwoven wood-pulp/polyester, flame-retardant, liquid repellant fabric
- Protect and preserve primary flame-resistant clothing
- Disposable
Can be used in steel mills, coal mines, maintenance, refineries, utilities, laboratories
Compliance
Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
Specifications
Medium | |
Blue | |
Elastic | |
Hooded | |
Zipper | |
Non-woven Wood Pulp, Polyester Fabric | |
28 in. | |
71.12 cm | |
33.75 in. | |
24.25 in. | |
1.6 to 1.73 m |
Tempro™ | |
Single-use | |
Elastic | |
Standard | |
Serged | |
35.25 to 38.75 in. | |
89.53 to 98.42 cm | |
Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities | |
85.72 cm | |
5.3 to 5.7 ft. |