missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Terephthaloyl Chloride 99.0+%, TCI America™
$31.14 - $98.62
Chemical Identifiers
CAS | 100-20-9 |
---|---|
Molecular Formula | C8H4Cl2O2 |
Molecular Weight (g/mol) | 203.02 |
MDL Number | MFCD00000693 |
InChI Key | LXEJRKJRKIFVNY-UHFFFAOYSA-N |
Synonym | terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride |
PubChem CID | 7488 |
IUPAC Name | benzene-1,4-dicarbonyl dichloride |
SMILES | ClC(=O)C1=CC=C(C=C1)C(Cl)=O |
Chemical Identifiers
100-20-9 | |
203.02 | |
LXEJRKJRKIFVNY-UHFFFAOYSA-N | |
7488 | |
ClC(=O)C1=CC=C(C=C1)C(Cl)=O |
C8H4Cl2O2 | |
MFCD00000693 | |
terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride | |
benzene-1,4-dicarbonyl dichloride |
Specifications
100-20-9 | |
White | |
C8H4Cl2O2 | |
25 g | |
terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride | |
ClC(=O)C1=CC=C(C=C1)C(Cl)=O | |
203.02 | |
203.02 | |
Crystalline Powder |
83°C | |
266°C | |
MFCD00000693 | |
2923 | |
LXEJRKJRKIFVNY-UHFFFAOYSA-N | |
benzene-1,4-dicarbonyl dichloride | |
7488 | |
≥99.0% (GC,T) | |
Terephthaloyl Chloride |
Safety and Handling
EINECSNumber : (3)-1378
RTECSNumber : WZ1797000
TSCA : Yes