missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Terephthaloyl Chloride 99.0+%, TCI America™
Supplier: TCI America T0017100G
Specifications
Terephthaloyl Chloride | |
83°C | |
266°C | |
MFCD00000693 | |
2923 | |
LXEJRKJRKIFVNY-UHFFFAOYSA-N | |
benzene-1,4-dicarbonyl dichloride | |
7488 | |
≥99.0% (GC,T) |
100-20-9 | |
White | |
C8H4Cl2O2 | |
100 g | |
terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride | |
ClC(=O)C1=CC=C(C=C1)C(Cl)=O | |
203.02 | |
203.02 | |
Crystalline Powder |
Chemical Identifiers
100-20-9 | |
203.02 | |
LXEJRKJRKIFVNY-UHFFFAOYSA-N | |
7488 | |
ClC(=O)C1=CC=C(C=C1)C(Cl)=O |
C8H4Cl2O2 | |
MFCD00000693 | |
terephthaloyl chloride, terephthaloyl dichloride, 1,4-benzenedicarbonyl dichloride, terephthalic acid dichloride, terephthalic dichloride, p-phthaloyl chloride, terephthalic acid chloride, p-phthalyl dichloride, p-phthaloyl dichloride, terephthalyl dichloride | |
benzene-1,4-dicarbonyl dichloride |
Safety and Handling
EINECSNumber : (3)-1378
RTECSNumber : WZ1797000
TSCA : Yes