Tergitol(R) 15-S-7, Surfactant, Spectrum™ Chemical
T1258, 84133-50-6, C12-14H25-29O[CH2CH2O]xH
Supplier:  Spectrum Chemical Mfg Cor T1258100ML
 View more versions of this product
Catalog No. 18607254
Explore available promotions
Description
Description
Spectrum™ Chemical Tergitol(R) 15-S-7, Surfactant is a Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.
Specifications
Specifications
5.5 to 7.5
C12-14-H25-29[CheCheO]xH
100 mL
Product Suggestions
Product Suggestions