Tergitol(R) 15-S-7, Surfactant, Spectrum™ Chemical
T1258, 84133-50-6, C12-14H25-29O[CH2CH2O]xH
$198.35 - $198.35
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | ||||||
18607254
|
Spectrum Chemical Mfg Cor
T1258100ML |
100 mL |
Each for $198.35
|
|
||||||
Description
Spectrum™ Chemical Tergitol(R) 15-S-7, Surfactant is a Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.Specifications
5.5 to 7.5 | |
C12-14-H25-29[CheCheO]xH |