missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Terpinene, 90%, Tech., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 207491000

 View more versions of this product

Catalog No. AC207491000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

α-Terpinene
99-86-5
100.0
0.8370g/mL
46°C
88% min. (GC)
C10H16
MFCD00001534
05, 126
14, 9170
Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether
CC(C)C1=CC=C(C)CC1
136.24
CHEBI:10334
90%
Liquid
tech., 90%
88.0
Colorless to Yellow
173°C to 175°C
Authentic
Glass bottle
1.4770 to 1.479
100 mL
0.837
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
YHQGMYUVUMAZJR-UHFFFAOYSA-N
1-methyl-4-propan-2-ylcyclohexa-1,3-diene
7462
136.24
Technical

Chemical Identifiers

99-86-5
136.24
YHQGMYUVUMAZJR-UHFFFAOYSA-N
7462
1-methyl-4-propan-2-ylcyclohexa-1,3-diene
C10H16
MFCD00001534
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
CHEBI:10334
CC(C)C1=CC=C(C)CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life with long last

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger

EINECSNumber : 202-795-1

RTECSNumber : OS8060000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only