Learn More
α-Terpinene, 90%, Tech., Thermo Scientific Chemicals
$60.60 - $248.98
Chemical Identifiers
CAS | 99-86-5 |
---|---|
Molecular Formula | C10H16 |
Molecular Weight (g/mol) | 136.24 |
MDL Number | MFCD00001534 |
InChI Key | YHQGMYUVUMAZJR-UHFFFAOYSA-N |
Synonym | alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene |
PubChem CID | 7462 |
ChEBI | CHEBI:10334 |
IUPAC Name | 1-methyl-4-propan-2-ylcyclohexa-1,3-diene |
SMILES | CC(C)C1=CC=C(C)CC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC207491000
|
Thermo Scientific Chemicals
207491000 |
100 mL | Glass bottle |
Each for $60.60
|
|
|||||
AC207495000
|
Thermo Scientific Chemicals
207495000 |
500 mL | Glass bottle |
Each for $248.98
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-86-5 | |
136.24 | |
YHQGMYUVUMAZJR-UHFFFAOYSA-N | |
7462 | |
1-methyl-4-propan-2-ylcyclohexa-1,3-diene |
C10H16 | |
MFCD00001534 | |
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene | |
CHEBI:10334 | |
CC(C)C1=CC=C(C)CC1 |
Specifications
99-86-5 | |
100.0 | |
0.8370g/mL | |
46°C | |
88% min. (GC) | |
C10H16 | |
MFCD00001534 | |
05, 126 | |
14, 9170 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether | |
CC(C)C1=CC=C(C)CC1 | |
136.24 | |
CHEBI:10334 | |
90% | |
Liquid |
88.0 | |
Colorless to Yellow | |
173°C to 175°C | |
Authentic | |
Glass bottle | |
1.4770 to 1.479 | |
100 mL | |
0.837 | |
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene | |
YHQGMYUVUMAZJR-UHFFFAOYSA-N | |
1-methyl-4-propan-2-ylcyclohexa-1,3-diene | |
7462 | |
136.24 | |
Technical | |
α-Terpinene, tech., 90% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life with long last
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha
GHS Signal Word: Danger
EINECSNumber : 202-795-1
RTECSNumber : OS8060000
TSCA : TSCA
RUO – Research Use Only