Learn More
gamma-Terpinene, 97%, stabilized
CAS: 99-85-4 | C10H16 | 136.24 g/mol
Supplier: Thermo Scientific Chemicals 207501000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| γ-Terpinene | |
| 99-85-4,10191-41-0 | |
| 0.8470g/mL | |
| 51°C | |
| 96% min. (GC) | |
| C10H16 | |
| MFCD00001537 | |
| 05, 128 | |
| 14, 9170 | |
| Solubility in water: less than 1%. Other solubilities: miscible with alcohol,ether | |
| CC1=CCC(=CC1)C(C)C | |
| 136.24 | |
| 7461 | |
| 136.24 | |
| Crystals or Powder |
| Stabilized | |
| White | |
| 182°C | |
| Authentic | |
| Glass bottle | |
| 1.4720 to 1.477 | |
| 100 mL | |
| 0.847 | |
| gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen | |
| YKFLAYDHMOASIY-UHFFFAOYSA-N | |
| 1-methyl-4-propan-2-ylcyclohexa-1,4-diene | |
| 0.1 % max. dl-alpha tocopherol | |
| CHEBI:10577 | |
| 97% |
Chemical Identifiers
| 99-85-4 | |
| 136.24 | |
| YKFLAYDHMOASIY-UHFFFAOYSA-N | |
| 7461 | |
| 1-methyl-4-propan-2-ylcyclohexa-1,4-diene |
| C10H16 | |
| MFCD00001537 | |
| gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen | |
| CHEBI:10577 | |
| CC1=CCC(=CC1)C(C)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for brea
GHS Signal Word: Danger
EINECSNumber : 202-794-6
RUO – Research Use Only