missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

γ-Terpinene, 97%, stabilized, Thermo Scientific Chemicals

$111.37 - $458.89

Chemical Identifiers

CAS 99-85-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001537
InChI Key YKFLAYDHMOASIY-UHFFFAOYSA-N
Synonym gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen
PubChem CID 7461
ChEBI CHEBI:10577
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,4-diene
SMILES CC1=CCC(=CC1)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC207501000
SDS View Documents
Thermo Scientific Chemicals
207501000
100 mL Glass bottle
Each for $111.37
 
AC207505000
SDS View Documents
Thermo Scientific Chemicals
207505000
500 mL Glass bottle
Each for $458.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99-85-4
136.24
YKFLAYDHMOASIY-UHFFFAOYSA-N
7461
1-methyl-4-propan-2-ylcyclohexa-1,4-diene
C10H16
MFCD00001537
gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen
CHEBI:10577
CC1=CCC(=CC1)C(C)C

Specifications

99-85-4,10191-41-0
0.8470g/mL
51°C
96% min. (GC)
C10H16
MFCD00001537
05, 128
14, 9170
Solubility in water: less than 1%. Other solubilities: miscible with alcohol,ether
CC1=CCC(=CC1)C(C)C
136.24
7461
136.24
Crystals or Powder
White
182°C
Authentic
Glass bottle
1.4720 to 1.477
100 mL
0.847
gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen
YKFLAYDHMOASIY-UHFFFAOYSA-N
1-methyl-4-propan-2-ylcyclohexa-1,4-diene
0.1 % max. dl-alpha tocopherol
CHEBI:10577
97%
γ-Terpinene, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for brea

GHS Signal Word: Danger

EINECSNumber : 202-794-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only