missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 1-piperazinecarboxylate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 367280050

 View more versions of this product

Catalog No. AC367280050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butyl 1-piperazinecarboxylate
43°C to 47°C
>110°C
96% min. (GC)
C9H19N2O2
5 g
1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1CC[NH2+]CC1
187.26
186.25
Crystalline Low Melting Solid
57260-71-6
White to Yellow
Authentic
Glass bottle
MFCD00075265
23, V,1, 55
CWXPZXBSDSIRCS-UHFFFAOYSA-O
tert-butyl piperazine-1-carboxylate
143452
97%

Chemical Identifiers

57260-71-6
187.26
CWXPZXBSDSIRCS-UHFFFAOYSA-O
143452
CC(C)(C)OC(=O)N1CC[NH2+]CC1
C9H19N2O2
MFCD00075265
1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
tert-butyl piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only