Learn More
tert-Butyl 1-piperazinecarboxylate, 97%, Thermo Scientific Chemicals
$144.85 - $144.85
Chemical Identifiers
CAS | 57260-71-6 |
---|---|
Molecular Formula | C9H19N2O2 |
Molecular Weight (g/mol) | 187.26 |
MDL Number | MFCD00075265 |
InChI Key | CWXPZXBSDSIRCS-UHFFFAOYSA-O |
Synonym | 1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester |
PubChem CID | 143452 |
IUPAC Name | tert-butyl piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC[NH2+]CC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC367280050
|
Thermo Scientific Chemicals
367280050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57260-71-6 | |
187.26 | |
CWXPZXBSDSIRCS-UHFFFAOYSA-O | |
143452 | |
CC(C)(C)OC(=O)N1CC[NH2+]CC1 |
C9H19N2O2 | |
MFCD00075265 | |
1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester | |
tert-butyl piperazine-1-carboxylate |
Specifications
57260-71-6 | |
White to Yellow | |
Authentic | |
Glass bottle | |
MFCD00075265 | |
23, V,1, 55 | |
CWXPZXBSDSIRCS-UHFFFAOYSA-O | |
tert-butyl piperazine-1-carboxylate | |
143452 | |
97% | |
tert-Butyl 1-piperazinecarboxylate |
43°C to 47°C | |
>110°C | |
96% min. (GC) | |
C9H19N2O2 | |
5 g | |
1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)N1CC[NH2+]CC1 | |
187.26 | |
186.25 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu
GHS Signal Word: Warning
RUO – Research Use Only