missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl acetoacetate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 107781000

 View more versions of this product

Catalog No. AC107781000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

tert-Butyl acetoacetate
1694-31-1
190°C
Authentic
Glass bottle
1.4190 to 1.421
MFCD00008811
01,83
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
JKUYRAMKJLMYLO-UHFFFAOYSA-N
tert-butyl 3-oxobutanoate
15538
97%
99%
0.9700g/mL
76°C
96% min. (GC)
C8H14O3
CH3COCH2CO2C(CH3)3
100mL
0.97
Solubility in water: 9g/L water (20°C). Other solubilities: soluble in xylene,miscible with most organic solvents
CC(=O)CC(=O)OC(C)(C)C
158.2
158.2

Chemical Identifiers

1694-31-1
158.2
JKUYRAMKJLMYLO-UHFFFAOYSA-N
15538
CC(=O)CC(=O)OC(C)(C)C
C8H14O3
MFCD00008811
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
tert-butyl 3-oxobutanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-904-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only