missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl acetoacetate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1694-31-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
MDL Number MFCD00008811
InChI Key JKUYRAMKJLMYLO-UHFFFAOYSA-N
Synonym tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
PubChem CID 15538
IUPAC Name tert-butyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC107781000
SDS View Documents
Thermo Scientific Chemicals
107781000
100mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC107785000
SDS View Documents
Thermo Scientific Chemicals
107785000
500mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1694-31-1
158.2
JKUYRAMKJLMYLO-UHFFFAOYSA-N
15538
CC(=O)CC(=O)OC(C)(C)C
C8H14O3
MFCD00008811
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
tert-butyl 3-oxobutanoate

Specifications

1694-31-1
190°C
Authentic
Glass bottle
1.4190 to 1.421
MFCD00008811
01,83
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
JKUYRAMKJLMYLO-UHFFFAOYSA-N
tert-butyl 3-oxobutanoate
15538
97%
0.9700g/mL
76°C
96% min. (GC)
C8H14O3
CH3COCH2CO2C(CH3)3
100mL
0.97
Solubility in water: 9g/L water (20°C). Other solubilities: soluble in xylene,miscible with most organic solvents
CC(=O)CC(=O)OC(C)(C)C
158.2
158.2
tert-Butyl acetoacetate, 99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-904-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only