missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 371131000

 View more versions of this product

Catalog No. AC371131000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butyl acrylate
1663-39-4,150-76-5
0.8830g/mL
11°C
98.5% min. (GC)
C7H12O2
H2C=CHCO2C(CH3)3
100 mL
0.883
Solubility in water: 2g/L
CC(C)(C)OC(=O)C=C
128.17
15458
128.17
Liquid
99%
-69.0°C
121.0°C
Authentic
Glass bottle
1.4100 to 1.4120
MFCD00008809
02, IV, 1465
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
tert-butyl prop-2-enoate
10 to 20ppm MEHQ
0.9 mPa.s (20°C)
99%

Chemical Identifiers

1663-39-4
128.17
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
15458
CC(C)(C)OC(=O)C=C
C7H12O2
MFCD00008809
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
tert-butyl prop-2-enoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

EINECSNumber : 216-768-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only