Learn More
tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
$107.56 - $215.44
Chemical Identifiers
CAS | 1663-39-4 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00008809 |
InChI Key | ISXSCDLOGDJUNJ-UHFFFAOYSA-N |
Synonym | tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester |
PubChem CID | 15458 |
IUPAC Name | tert-butyl prop-2-enoate |
SMILES | CC(C)(C)OC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC371131000
|
Thermo Scientific Chemicals
371131000 |
100 mL | Glass bottle |
Each for $107.56
|
|
||||
AC371130010
|
Thermo Scientific Chemicals
371130010 |
1 L | Glass bottle |
Each for $215.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1663-39-4 | |
128.17 | |
ISXSCDLOGDJUNJ-UHFFFAOYSA-N | |
15458 | |
CC(C)(C)OC(=O)C=C |
C7H12O2 | |
MFCD00008809 | |
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester | |
tert-butyl prop-2-enoate |
Specifications
1663-39-4,150-76-5 | |
0.8830g/mL | |
11°C | |
98.5% min. (GC) | |
C7H12O2 | |
H2C=CHCO2C(CH3)3 | |
100 mL | |
0.883 | |
Solubility in water: 2g/L | |
CC(C)(C)OC(=O)C=C | |
128.17 | |
15458 | |
128.17 | |
Liquid |
-69.0°C | |
121.0°C | |
Authentic | |
Glass bottle | |
1.4100 to 1.4120 | |
MFCD00008809 | |
02, IV, 1465 | |
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester | |
ISXSCDLOGDJUNJ-UHFFFAOYSA-N | |
tert-butyl prop-2-enoate | |
10 to 20ppm MEHQ | |
0.9 mPa.s (20°C) | |
99% | |
tert-Butyl acrylate, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 216-768-7
RUO – Research Use Only