missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl disulfide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 173622500

 View more versions of this product

Catalog No. AC173622500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butyl disulfide
-5.0°C
0.9300g/mL
67°C
97.5% min. (GC)
C8H18S2
[(CH3)3C]2S2
250 g
0.93
Solubility in water: insoluble
CC(C)(C)SSC(C)(C)C
178.35
178.35
Liquid
110-06-5
Yellow
198.0°C to 204.0°C
Authentic
Glass bottle
1.4882 to 1.4902
MFCD00008838
01,IV,1638
di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane
BKCNDTDWDGQHSD-UHFFFAOYSA-N
2-(tert-butyldisulfanyl)-2-methylpropane
66060
98%

Chemical Identifiers

110-06-5
178.35
BKCNDTDWDGQHSD-UHFFFAOYSA-N
66060
CC(C)(C)SSC(C)(C)C
C8H18S2
MFCD00008838
di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane
2-(tert-butyldisulfanyl)-2-methylpropane
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/

GHS Signal Word: Danger

EINECSNumber : 203-734-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only