Learn More
tert-Butyl disulfide, 98%, Thermo Scientific Chemicals
$290.15 - $714.37
Chemical Identifiers
CAS | 110-06-5 |
---|---|
Molecular Formula | C8H18S2 |
Molecular Weight (g/mol) | 178.35 |
MDL Number | MFCD00008838 |
InChI Key | BKCNDTDWDGQHSD-UHFFFAOYSA-N |
Synonym | di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane |
PubChem CID | 66060 |
IUPAC Name | 2-(tert-butyldisulfanyl)-2-methylpropane |
SMILES | CC(C)(C)SSC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC173622500
|
Thermo Scientific Chemicals
173622500 |
250 g | Glass bottle |
Each for $290.15
|
|
||||
AC173620010
|
Thermo Scientific Chemicals
173620010 |
1 kg | Glass bottle |
Each for $714.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-06-5 | |
178.35 | |
BKCNDTDWDGQHSD-UHFFFAOYSA-N | |
66060 | |
CC(C)(C)SSC(C)(C)C |
C8H18S2 | |
MFCD00008838 | |
di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane | |
2-(tert-butyldisulfanyl)-2-methylpropane |
Specifications
110-06-5 | |
Yellow | |
198.0°C to 204.0°C | |
Authentic | |
Glass bottle | |
1.4882 to 1.4902 | |
MFCD00008838 | |
01,IV,1638 | |
di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane | |
BKCNDTDWDGQHSD-UHFFFAOYSA-N | |
2-(tert-butyldisulfanyl)-2-methylpropane | |
66060 | |
98% | |
tert-Butyl disulfide |
-5.0°C | |
0.9300g/mL | |
67°C | |
97.5% min. (GC) | |
C8H18S2 | |
[(CH3)3C]2S2 | |
250 g | |
0.93 | |
Solubility in water: insoluble | |
CC(C)(C)SSC(C)(C)C | |
178.35 | |
178.35 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/
GHS Signal Word: Danger
EINECSNumber : 203-734-1
RUO – Research Use Only