Learn More
tert-Butyl ethyl ether, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2294522
Description
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.
Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.
Specifications
tert-Butyl ethyl ether | |
-94°C | |
72°C to 73°C | |
C6H14O | |
MFCD00009225 | |
UN1179 | |
14,3774 | |
Miscible with alcohol,ethyl ether. Slightly miscible with water. | |
CCOC(C)(C)C | |
102.18 | |
102.18 |
637-92-3 | |
0.75 | |
−19°C (−2°F) | |
1.375 | |
100 g | |
1731469 | |
tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane | |
NUMQCACRALPSHD-UHFFFAOYSA-N | |
2-ethoxy-2-methylpropane | |
12512 | |
99% |
Chemical Identifiers
637-92-3 | |
102.18 | |
NUMQCACRALPSHD-UHFFFAOYSA-N | |
12512 | |
CCOC(C)(C)C |
C6H14O | |
MFCD00009225 | |
tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane | |
2-ethoxy-2-methylpropane |
Safety and Handling
GHS H Statement
H225-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H225-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BUTYL ETHER
EINECSNumber : 211-309-7
RTECSNumber : KN4730200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only