missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl ethyl ether, 99%, Thermo Scientific Chemicals

$130.95 - $1132.23

Chemical Identifiers

CAS 637-92-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009225
InChI Key NUMQCACRALPSHD-UHFFFAOYSA-N
Synonym tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
PubChem CID 12512
IUPAC Name 2-ethoxy-2-methylpropane
SMILES CCOC(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2294506
View Documents
Thermo Scientific Chemicals
B2294506
5 g
Each for $130.95
 
AAB2294514
View Documents
Thermo Scientific Chemicals
B2294514
25 g
Each for $350.63
 
AAB2294522
View Documents
Thermo Scientific Chemicals
B2294522
100 g
Each for $1,132.23
 
Description

Description

tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.
Specifications

Chemical Identifiers

637-92-3
102.18
NUMQCACRALPSHD-UHFFFAOYSA-N
12512
CCOC(C)(C)C
C6H14O
MFCD00009225
tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
2-ethoxy-2-methylpropane

Specifications

637-92-3
0.75
−19°C (−2°F)
1.375
5 g
1731469
tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
NUMQCACRALPSHD-UHFFFAOYSA-N
2-ethoxy-2-methylpropane
12512
99%
-94°C
72°C to 73°C
C6H14O
MFCD00009225
UN1179
14,3774
Miscible with alcohol,ethyl ether. Slightly miscible with water.
CCOC(C)(C)C
102.18
102.18
tert-Butyl ethyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H225-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BUTYL ETHER

EINECSNumber : 211-309-7

RTECSNumber : KN4730200

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only