missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butyl glycolate, 94%, Thermo Scientific Chemicals
$405.47 - $1,059.18
Chemical Identifiers
CAS | 50595-15-8 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD03452739 |
InChI Key | WINGEFIITRDOLJ-UHFFFAOYSA-N |
Synonym | tert-butyl glycolate, 2-tert-butyl glycolate, 2-t-butyl glycolate, glycolic acid tert-butyl ester, hydroxyacetic acid tert-butyl ester, acetic acid, hydroxy-, 1,1-dimethylethyl ester, tert-butyl hydroxyacetate, t-butyl 2-hydroxyacetate, tert-butyl2-hydroxyacetate |
PubChem CID | 11815394 |
IUPAC Name | tert-butyl 2-hydroxyacetate |
SMILES | CC(C)(C)OC(=O)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH35711MD
|
Thermo Scientific Chemicals
H35711MD |
250 mg |
N/A
|
|
|||||
AAH3571103
|
Thermo Scientific Chemicals
H3571103 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50595-15-8 | |
132.16 | |
WINGEFIITRDOLJ-UHFFFAOYSA-N | |
11815394 | |
CC(C)(C)OC(=O)CO |
C6H12O3 | |
MFCD03452739 | |
tert-butyl glycolate, 2-tert-butyl glycolate, 2-t-butyl glycolate, glycolic acid tert-butyl ester, hydroxyacetic acid tert-butyl ester, acetic acid, hydroxy-, 1,1-dimethylethyl ester, tert-butyl hydroxyacetate, t-butyl 2-hydroxyacetate, tert-butyl2-hydroxyacetate | |
tert-butyl 2-hydroxyacetate |
Specifications
50595-15-8 | |
C6H12O3 | |
MFCD03452739 | |
tert-butyl glycolate, 2-tert-butyl glycolate, 2-t-butyl glycolate, glycolic acid tert-butyl ester, hydroxyacetic acid tert-butyl ester, acetic acid, hydroxy-, 1,1-dimethylethyl ester, tert-butyl hydroxyacetate, t-butyl 2-hydroxyacetate, tert-butyl2-hydroxyacetate | |
CC(C)(C)OC(=O)CO | |
132.16 | |
132.16 | |
Low Melting Solid |
Colorless to Yellow | |
HOCH2CO2C(CH3)3 | |
250 mg | |
WINGEFIITRDOLJ-UHFFFAOYSA-N | |
tert-butyl 2-hydroxyacetate | |
11815394 | |
94% | |
tert-Butyl glycolate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only