Learn More
tert-Butyl isocyanide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 403600010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
tert-Butyl isocyanide | |
7188-38-7 | |
91.0°C | |
96% min. (GC) | |
C5H9N | |
(CH3)3CNC | |
1 g | |
0.73 | |
FAGLEPBREOXSAC-UHFFFAOYSA-N | |
2-isocyano-2-methylpropane | |
23577 | |
97% |
97% | |
0.7300g/mL | |
−2°C | |
Glass bottle | |
1.3740 to 1.3780 | |
MFCD00000002 | |
02,50; 09,82; 11,99; 12,95; 14,63 | |
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane | |
CC(C)(C)[N+]#[C-] | |
83.13 | |
83.13 |
Chemical Identifiers
7188-38-7 | |
83.13 | |
FAGLEPBREOXSAC-UHFFFAOYSA-N | |
23577 | |
CC(C)(C)[N+]#[C-] |
C5H9N | |
MFCD00000002 | |
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane | |
2-isocyano-2-methylpropane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Fatal if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 230-553-5
RTECSNumber : EQ7102500
RUO – Research Use Only