missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl isocyanide, 97%, Thermo Scientific Chemicals

$70.27 - $70.27

Chemical Identifiers

CAS 7188-38-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00000002
InChI Key FAGLEPBREOXSAC-UHFFFAOYSA-N
Synonym tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
PubChem CID 23577
IUPAC Name 2-isocyano-2-methylpropane
SMILES CC(C)(C)[N+]#[C-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC403600010
SDS View Documents
Thermo Scientific Chemicals
403600010
1 g Glass bottle
Each for $70.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7188-38-7
83.13
FAGLEPBREOXSAC-UHFFFAOYSA-N
23577
CC(C)(C)[N+]#[C-]
C5H9N
MFCD00000002
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
2-isocyano-2-methylpropane

Specifications

7188-38-7
91.0°C
96% min. (GC)
C5H9N
(CH3)3CNC
1 g
0.73
FAGLEPBREOXSAC-UHFFFAOYSA-N
2-isocyano-2-methylpropane
23577
97%
0.7300g/mL
−2°C
Glass bottle
1.3740 to 1.3780
MFCD00000002
02,50; 09,82; 11,99; 12,95; 14,63
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
CC(C)(C)[N+]#[C-]
83.13
83.13
tert-Butyl isocyanide, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Fatal if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 230-553-5

RTECSNumber : EQ7102500

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only