missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl nitrite, tech. 90%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1613514

 View more versions of this product

Catalog No. AAL1613514


Explore available promotions

Description

Description

tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

Solubility
Miscible with diethyl ether, chloroform, alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.

Notes
Light sensitive. Store in cool place. Incompatible with strong reducing agents, powdered metals and strong acids.
Specifications

Specifications

tert-Butyl nitrite
0.867
−23°C (−9°F)
C4H9NO2
MFCD00002055
UN2351
14,1583
Miscible with diethyl ether,chloroform,alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.
CC(C)(C)ON=O
103.121
103.12
Technical
540-80-7
61°C to 63°C
Sweet
1.369
25 g
1209339
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
IOGXOCVLYRDXLW-UHFFFAOYSA-N
tert-butyl nitrite
10906
90%

Chemical Identifiers

540-80-7
103.121
IOGXOCVLYRDXLW-UHFFFAOYSA-N
10906
CC(C)(C)ON=O
C4H9NO2
MFCD00002055
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
tert-butyl nitrite
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c

H225-H302+H332

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL NITRITES

EINECSNumber : 208-757-0

RTECSNumber : RA0802000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only