missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl nitrite, tech. 90%, Thermo Scientific Chemicals

$65.81 - $146.04

Chemical Identifiers

CAS 540-80-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002055
InChI Key IOGXOCVLYRDXLW-UHFFFAOYSA-N
Synonym t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
PubChem CID 10906
IUPAC Name tert-butyl nitrite
SMILES CC(C)(C)ON=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1613514
View Documents
Thermo Scientific Chemicals
L1613514
25 g
Each for $65.81
 
AAL1613522
View Documents
Thermo Scientific Chemicals
L1613522
100 g
Each for $146.04
 
Description

Description

tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

Solubility
Miscible with diethyl ether, chloroform, alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.

Notes
Light sensitive. Store in cool place. Incompatible with strong reducing agents, powdered metals and strong acids.
Specifications

Chemical Identifiers

540-80-7
103.121
IOGXOCVLYRDXLW-UHFFFAOYSA-N
10906
CC(C)(C)ON=O
C4H9NO2
MFCD00002055
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
tert-butyl nitrite

Specifications

540-80-7
61°C to 63°C
Sweet
1.369
25 g
1209339
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
IOGXOCVLYRDXLW-UHFFFAOYSA-N
tert-butyl nitrite
10906
90%
tert-Butyl nitrite
0.867
−23°C (−9°F)
C4H9NO2
MFCD00002055
UN2351
14,1583
Miscible with diethyl ether,chloroform,alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.
CC(C)(C)ON=O
103.121
103.12
Technical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c

H225-H302+H332

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL NITRITES

EINECSNumber : 208-757-0

RTECSNumber : RA0802000

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only